Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027346.1 | NP_081622.1 | 3 | 535 | ||
GenBank mRNA | AK010072.1 | - | 3 | 553 | ||
AK047078.1 | - | 3 | 555 | |||
AK140143.1 | - | 3 | 573 | |||
AK147942.1 | - | 3 | 571 | |||
AK167963.1 | - | 3 | 552 | |||
BC029182.1 | - | 3 | 535 | 4195338 |