Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166416.1 | NP_001159888.1 | 20 | 2535 | ||
NM_027347.3 | NP_081623.3 | 19 | 2517 | |||
GenBank mRNA | AF507918.1 | - | 20 | 2485 | ||
AK028545.1 | - | 20 | 2548 | |||
AK129312.1 | - | 19 | 2450 | |||
BC005508.1 | - | 3 | 380 | |||
BC040398.1 | - | 19 | 2451 | |||
BC050916.1 | - | 19 | 2456 | 6405418 |