Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027354.2 | NP_081630.2 | 11 | 1522 | ||
GenBank mRNA | AK011064.1 | - | 10 | 1056 | ||
AK081217.1 | - | 707 | ||||
AK154163.1 | - | 11 | 1272 | |||
BC034901.1 | - | 11 | 1182 | 5255069 | ||
BE981233.1 | - | 1 | 184 |