Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146252.2 | NP_666364.1 | 10 | 1047 | ||
GenBank mRNA | AK158066.1 | - | 10 | 1128 | ||
BC025586.1 | - | 10 | 1073 | 5324875 | ||
BC039215.1 | - | 4 | 636 |