Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 14 | 2226 | ||
GenBank mRNA | AK145135.1 | - | 14 | 2112 | ||
AK172920.1 | - | 14 | 2093 | |||
BC071239.1 | - | 7 | 854 | |||
BC141056.1 | - | 14 | 2186 | 9055668 | ||
CB528069.1 | - | 4 | 456 |