Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 8 | 1128 | ||
GenBank mRNA | AK011138.1 | - | 8 | 1100 | ||
AK139553.1 | - | 8 | 1004 | |||
BC006674.1 | - | 8 | 1111 | 3499487 |