Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290984.1 | NP_001277913.1 | 5 | 536 | ||
NM_133728.3 | NP_598489.2 | 5 | 1967 | |||
GenBank mRNA | AK031295.1 | - | 5 | 1925 | ||
AK034746.1 | - | 1886 | ||||
AK049194.1 | - | 5 | 1914 | |||
AK050045.1 | - | 5 | 1907 | |||
AK076793.1 | - | 5 | 1869 | |||
AK155030.1 | - | 5 | 1906 | |||
AK168103.1 | - | 5 | 1907 | |||
AK168534.1 | - | 5 | 537 | |||
AK170510.1 | - | 5 | 1883 | |||
BC013617.1 | - | 5 | 1787 | 3979452 |