Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810408A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027419.3 | NP_081695.2 | 3 | 812 | ||
GenBank mRNA | AK013042.1 | - | 3 | 813 | ||
AK015537.1 | - | 3 | 738 | |||
AK051029.1 | - | 3 | 900 | |||
AK160144.1 | - | 3 | 722 | |||
AK171674.1 | - | 3 | 859 | |||
BC005538.1 | - | 2 | 104 | |||
BC051546.1 | - | 3 | 773 | 5255109 | ||
BC055710.1 | - | 3 | 917 | |||
BC069874.1 | - | 3 | 812 | 4216824 |