Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810408A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027419.3 | NP_081695.2 | 6 | 1274 | ||
GenBank mRNA | AK013042.1 | - | 6 | 1275 | ||
AK015537.1 | - | 6 | 1200 | |||
AK051029.1 | - | 6 | 1362 | |||
AK160144.1 | - | 6 | 1169 | |||
AK171674.1 | - | 6 | 1321 | |||
BC005538.1 | - | 5 | 566 | |||
BC051546.1 | - | 6 | 1236 | 5255109 | ||
BC055710.1 | - | 6 | 1379 | |||
BC069874.1 | - | 6 | 1274 | 4216824 |