Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 10 | 1059 | ||
GenBank mRNA | AK032897.1 | - | 8 | 598 | ||
AK048488.1 | - | 10 | 1082 | |||
AK050391.1 | - | 10 | 1059 | |||
AK078856.1 | - | 11 | 1018 | |||
AK131608.1 | - | 9 | 970 | |||
AK161812.1 | - | 9 | 1517 | |||
AK163839.1 | - | 1054 | ||||
AK168006.1 | - | 10 | 1060 | |||
AK168752.1 | - | 10 | 1081 | |||
AK169128.1 | - | 10 | 1060 | |||
AK214407.1 | - | 1 | 63 | |||
BC005755.1 | - | 10 | 1014 | 2654151 |