Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 7 | 907 | ||
GenBank mRNA | AK017523.1 | - | 8 | 1343 | ||
AK030908.1 | - | 2 | 2092 | |||
AK045166.1 | - | 1 | 107 | |||
AK075579.1 | - | 5 | 1207 | |||
AK154071.1 | - | 7 | 895 | |||
BC010777.1 | - | 7 | 907 | 4241357 |