Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 4 | 584 | ||
GenBank mRNA | AK017523.1 | - | 5 | 1020 | ||
AK075579.1 | - | 4 | 613 | |||
AK154071.1 | - | 4 | 572 | |||
BC010777.1 | - | 4 | 584 | 4241357 |