Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 9 | 989 | ||
GenBank mRNA | AK017582.1 | - | 9 | 884 | ||
AK034025.1 | - | 9 | 978 | |||
AK039945.1 | - | 9 | 856 | |||
AK046466.1 | - | 9 | 990 | |||
AK173153.1 | - | 1486 | ||||
BC052488.1 | - | 9 | 865 | 6438726 |