Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 15, 16 | 1780 | ||
GenBank mRNA | AK046466.1 | - | 15, 16 | 1781 | ||
AK173153.1 | - | 2277 | ||||
BC052488.1 | - | 15, 16 | 1656 | 6438726 |