Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081075.1 | NP_001074544.1 | 4, 5 | 379 | ||
GenBank mRNA | AK017666.1 | - | 4, 5 | 441 | ||
AK077572.1 | - | 4, 5 | 352 | |||
AK172241.1 | - | 4, 5 | 386 |