Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276767.1 | NP_001263696.1 | 8 | 1017 | ||
NM_144530.6 | NP_653113.4 | 7 | 963 | |||
GenBank mRNA | AK041148.1 | - | 7 | 937 | ||
AK078105.1 | - | 7 | 926 | |||
AK122341.2 | - | 6 | 4498 | |||
AK134057.1 | - | 4 | 446 | |||
AK145740.1 | - | 7 | 936 | |||
AK167700.1 | - | 7 | 937 | |||
BC005786.1 | - | 6 | 780 | 2811511 | ||
BC058552.1 | - | 6 | 1022 | |||
BC066163.1 | - | 8 | 975 | 30092453 | ||
BC066848.1 | - | 6 | 1774 | 30471872 | ||
FR682665.1 | - | 8 | 936 |
Gene Symbol: Zbed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FR682664.1 | - | 6775 |