Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159581.1 | NP_001153053.1 | 6, 7 | 515 | ||
NM_133250.2 | NP_573513.2 | 5, 6 | 422 | |||
GenBank mRNA | AB117938.1 | - | 4, 5 | 384 | ||
AB117939.1 | - | 6, 7 | 499 | |||
AK017730.1 | - | 5, 6 | 410 | |||
AK077546.1 | - | 5, 6 | 419 | |||
AK156790.1 | - | 6, 7 | 505 | |||
AY007717.1 | - | 4, 5 | 378 | |||
BC057942.1 | - | 5, 6 | 403 | 5320744 |