Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 17, 18 | 2265 | ||
GenBank mRNA | AK129257.1 | - | 11, 12 | 1278 | ||
BC063268.1 | - | 17, 18 | 2061 | 4187769 | ||
BC080688.1 | - | 17, 18 | 2207 | 6515742 | ||
BC082313.1 | - | 3, 4 | 420 |