Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177995.1 | NP_001171466.1 | 16 | 3706 | ||
NM_001291026.1 | NP_001277955.1 | 17 | 3875 | |||
NM_001291029.1 | NP_001277958.1 | 16 | 3709 | |||
NM_027504.3 | NP_081780.3 | 17 | 3878 | |||
GenBank mRNA | AB078338.1 | - | 17 | 3770 | ||
AK017846.1 | - | 7 | 940 | |||
AK083829.1 | - | 17 | 3766 | |||
AK152396.1 | - | 6 | 1007 | |||
AK153563.1 | - | 6 | 1008 | |||
AK173230.1 | - | 9 | 2060 | |||
BC053082.1 | - | 16 | 3640 | |||
BC059838.1 | - | 16 | 3687 | 6409778 |