Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030104.1 | - | 1 | 1348 | ||
AK122563.1 | - | 1 | 1361 | |||
AK144688.1 | - | 1 | 1371 | |||
BC006059.1 | - | 1 | 1333 | |||
BC059209.1 | - | 1 | 1367 | 6406239 |