Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028450.3 | NP_082726.2 | 8 | 818 | ||
GenBank mRNA | AK014093.1 | - | 7 | 567 | ||
AK017798.1 | - | 8 | 796 | |||
AK028490.1 | - | 7 | 734 | |||
AK036670.1 | - | 8 | 818 | |||
BC032154.1 | - | 8 | 989 | 5068339 | ||
BY713087.1 | - | 7 | 565 |