Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027533.3 | NP_081809.2 | 1 | 266 | ||
GenBank mRNA | AK018178.1 | - | 1 | 69 | ||
AK032520.1 | - | 1 | 92 | |||
AK049888.1 | - | 1 | 68 | |||
AK079560.1 | - | 1 | 70 | |||
BB642061.1 | - | 1 | 267 | |||
BC007185.1 | - | 1 | 507 | 3591472 |