Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081359.3 | NP_001074828.2 | 39 | 5750 | ||
NM_001112721.2 | NP_001106192.1 | 39 | 5732 | |||
GenBank mRNA | AK122398.1 | - | 16 | 2223 | ||
AY550908.1 | - | 39 | 5340 | |||
BC047348.1 | - | 30 | 4243 | |||
BC049162.1 | - | 10 | 1430 |