Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081359.3 | NP_001074828.2 | 47 | 7135 | ||
NM_001112721.2 | NP_001106192.1 | 47 | 7117 | |||
GenBank mRNA | AK122398.1 | - | 24 | 3608 | ||
AY550908.1 | - | 47 | 6725 | |||
BC047348.1 | - | 38 | 5628 | |||
BC049162.1 | - | 18 | 2815 |