Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081359.3 | NP_001074828.2 | 34 | 4956 | ||
NM_001112721.2 | NP_001106192.1 | 34 | 4938 | |||
GenBank mRNA | AK122398.1 | - | 11 | 1429 | ||
AY550908.1 | - | 34 | 4546 | |||
BC047348.1 | - | 25 | 3449 | |||
BC049162.1 | - | 5 | 636 | |||
CB527808.1 | - | 1 | 139 | |||
CK636472.1 | - | 2 | 288 |