Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003909.4 | NP_001003909.2 | 8 | 1785 | ||
NM_001289527.1 | NP_001276456.1 | 8 | 1785 | |||
NM_001289528.1 | NP_001276457.1 | 7 | 1667 | |||
NM_001289529.1 | NP_001276458.1 | 7 | 1257 | |||
GenBank mRNA | AK129343.1 | - | 8 | 1586 | ||
BC020475.1 | - | 8 | 1319 | |||
BC089031.1 | - | 7 | 1236 | |||
BC145510.1 | - | 7 | 1418 | |||
BC145511.1 | - | 8 | 1536 | 9054074 | ||
BC150792.1 | - | 8 | 1536 | 9087703 |