Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025428.1 | NP_001020599.1 | 13 | 1627 | ||
NM_001025429.1 | NP_001020600.1 | 13 | 1624 | |||
NM_001025430.1 | NP_001020601.1 | 12 | 1612 | |||
NM_001199203.1 | NP_001186132.1 | 16 | 2103 | |||
NM_001199204.1 | NP_001186133.1 | 15 | 2061 | |||
NM_001199205.1 | NP_001186134.1 | 12 | 1612 | |||
NM_027569.2 | NP_081845.2 | 15 | 2061 | |||
GenBank mRNA | AB047782.1 | - | 16 | 2023 | ||
AF327451.1 | - | 15 | 2043 | |||
AK014638.1 | - | 8 | 734 | |||
AK049161.1 | - | 6 | 624 | |||
AK147431.1 | - | 12 | 1669 | |||
AK147537.1 | - | 12 | 1688 | |||
AK172961.1 | - | 13 | 1606 | |||
AY823270.1 | - | 15 | 1605 | |||
BC052058.1 | - | 13 | 1562 | |||
BC060100.1 | - | 12 | 1546 | 6417257 | ||
BC094670.1 | - | 12 | 1540 | 30618738 | ||
FJ042496.1 | - | 15 | 1938 |