Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027572.1 | NP_081848.1 | 5, 6 | 1422 | ||
GenBank mRNA | AK014809.1 | - | 2, 3 | 655 | ||
BC100473.1 | - | 5, 6 | 1422 | 30918267 | ||
BC138326.1 | - | 5, 6 | 1433 | 8861346 | ||
BC138327.1 | - | 5, 6 | 1433 | 8861347 | ||
DQ164804.2 | - | 5, 6 | 1448 | |||
DQ164805.1 | - | 5, 6 | 1409 |