Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027572.1 | NP_081848.1 | 2 | 535 | ||
GenBank mRNA | BC100473.1 | - | 2 | 535 | 30918267 | |
BC138326.1 | - | 2 | 546 | 8861346 | ||
BC138327.1 | - | 2 | 546 | 8861347 | ||
DQ164804.2 | - | 2 | 561 | |||
DQ164805.1 | - | 2 | 522 |