Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027588.3 | NP_081864.2 | 2, 3 | 216 | ||
GenBank mRNA | AF356186.1 | - | 2, 3 | 189 | ||
AJ295253.1 | - | 2, 3 | 143 | |||
AK014892.1 | - | 2, 3 | 216 | |||
BB573272.1 | - | 2, 3 | 221 | |||
BC060976.1 | - | 2, 3 | 208 | 6773248 |