Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081046.1 | NP_001074515.1 | 3 | 393 | ||
GenBank mRNA | AK014861.1 | - | 3 | 387 | ||
AK162207.1 | - | 12 | 878 | |||
BC050801.1 | - | 3 | 391 | 6770929 |