Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162846.1 | NP_001156318.1 | 5 | 849 | ||
NM_178621.4 | NP_848736.2 | 5 | 999 | |||
GenBank mRNA | AK029522.1 | - | 5 | 986 | ||
AK032166.1 | - | 5 | 1000 | |||
AK034448.1 | - | 5 | 979 | |||
AK083011.1 | - | 5 | 1000 | |||
BC027081.1 | - | 3 | 439 | |||
BC037618.1 | - | 5 | 987 | 5400532 |