Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172407.3 | NP_765995.1 | 3 | 1087 | ||
GenBank mRNA | AK014874.1 | - | 2 | 632 | ||
AK048437.1 | - | 3 | 1056 | |||
AK078329.1 | - | 3 | 1054 | |||
AK089057.1 | - | 3 | 1057 | |||
AK133831.1 | - | 3 | 1088 | |||
AK167684.1 | - | 3 | 1055 | |||
AK202731.1 | - | 1 | 233 |