Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138757.2 | NP_620096.2 | 11 | 1778 | ||
GenBank mRNA | AK182579.1 | - | 2 | 227 | ||
BC026851.1 | - | 11 | 1707 | 4459471 | ||
BC027229.1 | - | 11 | 1363 | 2615769 | ||
CK618181.1 | - | 6 | 471 |