Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207301.2 | NP_997184.1 | 4 | 491 | ||
GenBank mRNA | AK021091.1 | - | 4 | 490 | ||
AK172079.1 | - | 4 | 504 | |||
BC031769.1 | - | 4 | 491 | 4235618 |