Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027810.3 | NP_082086.2 | 13, 14 | 1476 | ||
GenBank mRNA | AF521645.1 | - | 13, 14 | 1475 | ||
AK050291.1 | - | 13, 14 | 1474 | |||
AK132661.1 | - | 13, 14 | 1404 | |||
BC031505.1 | - | 13, 14 | 1426 | 3494587 |