Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB048364.1 | - | 11 | 1198 | ||
AK018634.1 | - | 11 | 1025 | |||
AK086508.1 | - | 11 | 1192 | |||
BC038274.1 | - | 11 | 1268 | 4457335 |