Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164477.1 | NP_001157949.1 | 13 | 2870 | ||
NM_027835.3 | NP_082111.2 | 14 | 3017 | |||
GenBank mRNA | AF374384.1 | - | 14 | 2926 | ||
AK018602.1 | - | 3 | 351 | |||
AK153018.1 | - | 14 | 2924 | |||
AK153344.1 | - | 6 | 951 | |||
AY075132.1 | - | 14 | 2692 | |||
BC004031.1 | - | 7 | 1159 | |||
BC019605.1 | - | 1 | 48 | |||
BC025508.1 | - | 9 | 1461 | |||
BC080200.1 | - | 13 | 2775 | 3597815 | ||
EF113547.1 | - | 14 | 2706 | |||
EF113548.1 | - | 14 | 2706 | |||
EU084750.1 | - | 14 | 2706 |