Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164477.1 | NP_001157949.1 | 1 | 698 | ||
NM_027835.3 | NP_082111.2 | 1 | 698 | |||
GenBank mRNA | AF374384.1 | - | 1 | 607 | ||
AK037057.1 | - | 1 | 596 | |||
AK040519.1 | - | 1 | 655 | |||
AK149802.1 | - | 1 | 605 | |||
AK153018.1 | - | 1 | 605 | |||
AK156450.1 | - | 1 | 606 | |||
AK168646.1 | - | 1 | 699 | |||
AY075132.1 | - | 1 | 373 | |||
BC080200.1 | - | 1 | 603 | 3597815 | ||
EF113547.1 | - | 1 | 387 | |||
EF113548.1 | - | 1 | 387 | |||
EU084750.1 | - | 1 | 387 |