Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF521095.1 | - | 5 | 1171 | ||
AK004878.1 | - | 5 | 1231 | |||
AK159405.1 | - | 5 | 1220 | |||
AK159912.1 | - | 4 | 1520 | |||
AK166894.1 | - | 4 | 1514 | |||
AK170330.1 | - | 4 | 1529 | |||
BC020046.1 | - | 4 | 641 | |||
BC064753.1 | - | 4 | 1159 |