Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023622.4 | NP_076111.2 | 13, 14 | 1540 | ||
GenBank mRNA | AF237669.1 | - | 13, 14 | 1499 | ||
AF239661.1 | - | 13, 14 | 1541 | |||
AK004876.1 | - | 10, 11 | 1569 | |||
AK053550.1 | - | 13, 14 | 1527 | |||
AK131140.1 | - | 1046 | ||||
AK134654.1 | - | 1522 | ||||
AK165023.1 | - | 10, 11 | 1929 | |||
AK165746.1 | - | 11, 12 | 1831 |