Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1300017J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027918.2 | NP_082194.1 | 10, 11 | 1339 | ||
GenBank mRNA | AK005035.1 | - | 10, 11 | 1340 | ||
BC021390.1 | - | 10, 11 | 1335 | 5054320 |