Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023627.1 | NP_076116.1 | 9 | 1302 | ||
GenBank mRNA | AF288525.1 | - | 9 | 1302 | ||
AK005029.2 | - | 9 | 1280 | |||
AK079323.1 | - | 9 | 1279 | |||
AK137089.1 | - | 8 | 1384 | |||
BC003458.1 | - | 9 | 1293 | 2648362 |