Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253814.1 | NP_001240743.1 | 12 | 2820 | ||
NM_027922.2 | NP_082198.1 | 12 | 2817 | |||
GenBank mRNA | AK003234.1 | - | 9 | 1619 | ||
AK129192.1 | - | 12 | 1929 | |||
AK132187.1 | - | 12 | 2822 | |||
AK134966.1 | - | 2 | 652 | |||
AK136587.1 | - | 2 | 240 | |||
AK142497.1 | - | 10 | 1869 | |||
BC065071.1 | - | 12 | 2780 | 6410033 |