Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170591.1 | NP_733479.1 | 5 | 726 | ||
GenBank mRNA | AK006062.1 | - | 5 | 828 | ||
AK029688.1 | - | 780 | ||||
AK078279.1 | - | 5 | 774 | |||
AK086412.1 | - | 5 | 794 | |||
AK129134.1 | - | 5 | 724 | |||
AK133721.1 | - | 5 | 737 | |||
AK139895.1 | - | 5 | 857 | |||
AK146477.1 | - | 5 | 726 | |||
AK163792.1 | - | 5 | 755 | |||
BC026743.1 | - | 5 | 757 | 3964978 |