Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027959.3 | NP_082235.1 | 6 | 564 | ||
GenBank mRNA | AK005989.1 | - | 6 | 570 | ||
AK076558.1 | - | 6 | 565 | |||
AK133792.1 | - | 6 | 571 | |||
AK151008.1 | - | 6 | 545 | |||
AK152960.1 | - | 6 | 577 | |||
AK165877.1 | - | 6 | 543 | |||
AK166111.1 | - | 6 | 571 | |||
AK167386.1 | - | 6 | 582 | |||
AK168136.1 | - | 6 | 571 | |||
AK168279.1 | - | 6 | 571 | |||
BC006865.1 | - | 6 | 553 | 2645183 |