Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284524.1 | NP_001271453.1 | 6 | 586 | ||
NM_001284525.1 | NP_001271454.1 | 6 | 622 | |||
NM_027979.2 | NP_082255.1 | 6 | 586 | |||
GenBank mRNA | AK009012.1 | - | 6 | 587 | ||
AY458654.1 | - | 6 | 586 | |||
AY536287.1 | - | 5 | 460 | |||
BC138765.1 | - | 6 | 588 | 8861787 |