Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027988.4 | NP_082264.2 | 6 | 1582 | ||
GenBank mRNA | AF399754.1 | - | 5 | 568 | ||
AF539797.1 | - | 5 | 568 | |||
AK009605.1 | - | 5 | 677 | |||
AK088226.1 | - | 6 | 1541 | |||
AK157993.1 | - | 6 | 1583 | |||
BC019525.1 | - | 5 | 594 | 4220018 | ||
BC119527.1 | - | 5 | 670 | |||
BC119528.1 | - | 3 | 899 | |||
BC127235.1 | - | 5 | 670 | |||
BC127236.1 | - | 5 | 670 |
Gene Symbol: Tbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137554.1 | - | 3698 |