Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162883.1 | NP_001156355.1 | 3 | 1012 | ||
NM_173786.1 | NP_776147.1 | 3 | 1064 | |||
GenBank mRNA | BC020489.1 | - | 3 | 1042 | 2649139 | |
BC057684.1 | - | 3 | 1072 | 3675306 |
Gene Symbol: Apol9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 1083 | ||
NM_173743.4 | NP_776104.1 | 3 | 1034 | |||
GenBank mRNA | AK009386.1 | - | 3 | 1085 | ||
AK050167.1 | - | 3 | 1036 | |||
BF148017.1 | - | 2 | 441 |