Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162883.1 | NP_001156355.1 | 3 | 361 | ||
NM_173786.1 | NP_776147.1 | 3 | 413 | |||
GenBank mRNA | AI530062.1 | - | 3 | 413 | ||
BC020489.1 | - | 3 | 391 | 2649139 | ||
BC057684.1 | - | 3 | 421 | 3675306 | ||
CJ058381.1 | - | 3 | 362 |
Gene Symbol: Apol9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 432 | ||
NM_173743.4 | NP_776104.1 | 3 | 383 | |||
GenBank mRNA | AK009386.1 | - | 3 | 433 | ||
AK050167.1 | - | 3 | 385 |